missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyldodecane, 97%

Catalog No. p-3779421
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC151020100 10 g Glass bottle
AC151020500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC151020100 Supplier Thermo Scientific Chemicals Supplier No. 151020100
Only null left

CAS: 123-01-3 | C18H30 | 246.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 123-01-3
Molecular Formula C18H30
Molecular Weight (g/mol) 246.43
MDL Number MFCD00008974
InChI Key KWKXNDCHNDYVRT-UHFFFAOYSA-N
Synonym 1-phenyldodecane, benzene, dodecyl, n-dodecylbenzene, laurylbenzene, phenyldodecane, marlican, dodecane, 1-phenyl, detergent alkylate, alkylate p 1, phenyldodecan
PubChem CID 31237
IUPAC Name dodecylbenzene
SMILES CCCCCCCCCCCCC1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -7°C
Density 0.8500g/mL
Boiling Point 288°C to 300°C
Flash Point 124°C
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.4810 to 1.483
Linear Formula C6H5(CH2)11CH3
Quantity 10 g
Specific Gravity 0.85
Solubility Information Solubility in water: immiscible with water
Formula Weight 246.43
Percent Purity 97%
Chemical Name or Material 1-Phenyldodecane
Show More Show Less
EINECSNumber 204-591-8

RUO – Research Use Only