missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylcyclopentanecarboxylic acid, 98%

Catalog No. AC161140050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC161140050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC161140050 Supplier Thermo Scientific Chemicals Supplier No. 161140050
Only null left

CAS: 77-55-4 | C12H14O2 | 190.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 77-55-4
Molecular Formula C12H14O2
Molecular Weight (g/mol) 190.24
MDL Number MFCD00001372
InChI Key RHPCYZLXNNRRMB-UHFFFAOYSA-N
Synonym 1-phenylcyclopentanecarboxylic acid, 1-phenyl-1-cyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 1-phenyl, 1-phenyl-cyclopentanecarboxylic acid, 1-phenylcyclopentane carboxylic acid, cyclopentanecarboxylicacid, 1-phenyl, pubchem22923, acmc-209pao, phenylcyclopentanecarboxylic acid, phenylcyclopentane carboxylic acid
PubChem CID 66167
IUPAC Name 1-phenylcyclopentane-1-carboxylic acid
SMILES OC(=O)C1(CCCC1)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 157°C to 161°C
Color Brown-Gray to Cream
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H5C5H8CO2H
Quantity 5 g
Beilstein 09, II, 418
Formula Weight 190.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1-Phenylcyclopentanecarboxylic acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-037-7
TSCA TSCA

RUO – Research Use Only