missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylcyclohexene, 96%

Catalog No. p-7025224
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2060206 5 g
AAB2060214 25 g
AAB2060222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2060206 Supplier Thermo Scientific Chemicals Supplier No. B2060206

CAS: 771-98-2 | C12H14 | 158.244 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 771-98-2
Molecular Formula C12H14
Molecular Weight (g/mol) 158.244
MDL Number MFCD00001542
InChI Key WCMSFBRREKZZFL-UHFFFAOYSA-N
Synonym 1-phenyl-1-cyclohexene, 1-phenylcyclohexene, 1-phenylcyclohex-1-ene, benzene, 1-cyclohexen-1-yl, phenylcyclohexene, cyclohexenylbenzene, benzene, cyclohexenyl, 2,3,4,5-tetrahydro-1,1'-biphenyl, cyclohexene, 1-phenyl, trans-1-phenylcyclohexene
PubChem CID 13043
IUPAC Name cyclohexen-1-ylbenzene
SMILES C1CCC(=CC1)C2=CC=CC=C2

Specifications

Melting Point -11°C
Density 0.992
Boiling Point 250°C to 252°C
Flash Point 103°C (217°F)
Refractive Index 1.57
Quantity 5 g
Beilstein 1905772
Solubility Information Insoluble in water.
Formula Weight 158.24
Percent Purity 97%
Chemical Name or Material 1-Phenylcyclohexene
Show More Show Less
EINECSNumber 212-242-6
RTECSNumber GW6600000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only