missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylcyclohexanol 98.0+%, TCI America™

Catalog No. P19885G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P19885G 5 g
P198825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P19885G Supplier TCI America Supplier No. P19885G

Chemical Identifiers

CAS 1589-60-2
Molecular Formula C12H16O
Molecular Weight (g/mol) 176.259
MDL Number MFCD00021393
InChI Key DTTDXHDYTWQDCS-UHFFFAOYSA-N
Synonym 1-phenylcyclohexanol, cyclohexanol, 1-phenyl, 1-phenyl-1-cyclohexanol, phenylcyclohexanol, cyclohexanol, phenyl, 1-phenylcyclohexanol-1, 1-phenyl-cyclohexanol, maybridge1_002485, 1-phenyl-cyclohexan-1-ol, acmc-209dj8
PubChem CID 15319
IUPAC Name 1-phenylcyclohexan-1-ol
SMILES C1CCC(CC1)(C2=CC=CC=C2)O

Specifications

Melting Point 63°C
Color White-Yellow
Boiling Point 100°C
Quantity 5 g
Formula Weight 176.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Phenylcyclohexanol
RTECSNumber GW0699900
TSCA No