missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylcyclohexanol, 97%

Catalog No. AAB2087718
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2087718 50 g
AAB2087709 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2087718 Supplier Thermo Scientific Chemicals Supplier No. B2087718
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1589-60-2 | C12H16O | 176.259 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1589-60-2
Molecular Formula C12H16O
Molecular Weight (g/mol) 176.259
MDL Number MFCD00021393
InChI Key DTTDXHDYTWQDCS-UHFFFAOYSA-N
Synonym 1-phenylcyclohexanol, cyclohexanol, 1-phenyl, 1-phenyl-1-cyclohexanol, phenylcyclohexanol, cyclohexanol, phenyl, 1-phenylcyclohexanol-1, 1-phenyl-cyclohexanol, maybridge1_002485, 1-phenyl-cyclohexan-1-ol, acmc-209dj8
PubChem CID 15319
IUPAC Name 1-phenylcyclohexan-1-ol
SMILES C1CCC(CC1)(C2=CC=CC=C2)O

Specifications

Melting Point 58°C to 62°C
Boiling Point 153°C (20 mmHg)
Quantity 50 g
Beilstein 1818360
Formula Weight 176.26
Percent Purity 97%
Chemical Name or Material 1-Phenylcyclohexanol
EINECSNumber 216-456-0
RTECSNumber GW0699900
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only