missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-3-guanylthiourea, TCI America™

Catalog No. P11675G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P11675G Supplier TCI America Supplier No. P11675G
Only null left

Chemical Identifiers

CAS 15989-47-6
Molecular Formula C8H10N4S
Molecular Weight (g/mol) 194.256
MDL Number MFCD00191480
InChI Key IKGVILAOZDBXSV-UHFFFAOYSA-N
Synonym 1-Amidino-3-phenylthiourea, 1-Carbamimidoyl-3-phenylthiourea
PubChem CID 5181676
IUPAC Name 1-(diaminomethylidene)-3-phenylthiourea
SMILES C1=CC=C(C=C1)NC(=S)N=C(N)N

Specifications

Melting Point 169°C
Color White-Yellow
Quantity 5 g
Formula Weight 194.26
Physical Form Crystalline Powder
Chemical Name or Material 1-Phenyl-3-guanylthiourea