missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Phenyl-2-(trimethylsilyl)acetylene 98.0+%, TCI America™
$96.26 - $354.23
Chemical Identifiers
| CAS | 2170-06-1 |
|---|---|
| Molecular Formula | C11H14Si |
| Molecular Weight (g/mol) | 174.32 |
| MDL Number | MFCD00054860 |
| InChI Key | UZIXCCMXZQWTPB-UHFFFAOYSA-N |
| Synonym | 1-phenyl-2-trimethylsilyl acetylene, trimethyl phenylethynyl silane, silane, trimethyl phenylethynyl, 1-phenyl-2-trimethylsilylacetylene, phenylethynyltrimethylsilane, trimethylsilyl phenylacetylene, trimethyl 2-phenylethynyl silane, phenylethynyl-trimethylsilane, phenylethynyl trimethylsilane, acmc-1bika |
| PubChem CID | 137464 |
| IUPAC Name | trimethyl(2-phenylethynyl)silane |
| SMILES | C[Si](C)(C)C#CC1=CC=CC=C1 |
Chemical Identifiers
| 2170-06-1 | |
| 174.32 | |
| UZIXCCMXZQWTPB-UHFFFAOYSA-N | |
| 137464 | |
| C[Si](C)(C)C#CC1=CC=CC=C1 |
| C11H14Si | |
| MFCD00054860 | |
| 1-phenyl-2-trimethylsilyl acetylene, trimethyl phenylethynyl silane, silane, trimethyl phenylethynyl, 1-phenyl-2-trimethylsilylacetylene, phenylethynyltrimethylsilane, trimethylsilyl phenylacetylene, trimethyl 2-phenylethynyl silane, phenylethynyl-trimethylsilane, phenylethynyl trimethylsilane, acmc-1bika | |
| trimethyl(2-phenylethynyl)silane |
Specifications
| 2170-06-1 | |
| 88°C | |
| MFCD00054860 | |
| 1-phenyl-2-trimethylsilyl acetylene, trimethyl phenylethynyl silane, silane, trimethyl phenylethynyl, 1-phenyl-2-trimethylsilylacetylene, phenylethynyltrimethylsilane, trimethylsilyl phenylacetylene, trimethyl 2-phenylethynyl silane, phenylethynyl-trimethylsilane, phenylethynyl trimethylsilane, acmc-1bika | |
| C[Si](C)(C)C#CC1=CC=CC=C1 | |
| 174.32 | |
| 174.32 | |
| Liquid |
| Yellow | |
| C11H14Si | |
| 5 mL | |
| UZIXCCMXZQWTPB-UHFFFAOYSA-N | |
| trimethyl(2-phenylethynyl)silane | |
| 137464 | |
| ≥98.0% (GC) | |
| 1-Phenyl-2-(trimethylsilyl)acetylene |
Safety and Handling
TSCA : No