missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-2-propyn-1-ol 98.0+%, TCI America™

Catalog No. P02205G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P02205G 5 g
P022025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P02205G Supplier TCI America Supplier No. P02205G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4187-87-5
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.162
MDL Number MFCD00021860
InChI Key UIGLAZDLBZDVBL-UHFFFAOYSA-N
Synonym 1-phenyl-2-propyn-1-ol, 1-phenylpropargyl alcohol, ethynylphenylcarbinol, phenylethynylcarbinol, 2-propyn-1-ol, 1-phenyl, benzyl alcohol, .alpha.-ethynyl, benzenemethanol, .alpha.-ethynyl, alpha-ethynylbenzyl alcohol, alpha-ethynylbenzenemethanol, alpha-phenylpropargyl alcohol
PubChem CID 20155
IUPAC Name 1-phenylprop-2-yn-1-ol
SMILES C#CC(C1=CC=CC=C1)O

Specifications

Color White-Yellow
Boiling Point 115°C
Quantity 5 g
Formula Weight 132.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Phenyl-2-propyn-1-ol
RTECSNumber DO5900000
TSCA No