missing translation for 'onlineSavingsMsg'
Learn More

(±)-1-Phenyl-2-propyn-1-ol, 98+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 392760010

 View more versions of this product

Catalog No. AC392760010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

(±)-1-Phenyl-2-propyn-1-ol
98.0
1.0660g/mL
99°C
98% min. (GC)
C9H8O
HC≡CCH(OH)C6H5
1g
1-phenyl-2-propyn-1-ol, 1-phenylpropargyl alcohol, ethynylphenylcarbinol, phenylethynylcarbinol, 2-propyn-1-ol, 1-phenyl, benzyl alcohol, .alpha.-ethynyl, benzenemethanol, .alpha.-ethynyl, alpha-ethynylbenzyl alcohol, alpha-ethynylbenzenemethanol, alpha-phenylpropargyl alcohol
C#CC(C1=CC=CC=C1)O
132.16
132.16
4187-87-5
100.0
231.0°C to 233.0°C
Authentic
Glass bottle
1.5480 to 1.5510 (20°C, 589nm)
MFCD00021860
1.066
UIGLAZDLBZDVBL-UHFFFAOYSA-N
1-phenylprop-2-yn-1-ol
20155
98+%

Chemical Identifiers

4187-87-5
132.16
UIGLAZDLBZDVBL-UHFFFAOYSA-N
20155
C#CC(C1=CC=CC=C1)O
C9H8O
MFCD00021860
1-phenyl-2-propyn-1-ol, 1-phenylpropargyl alcohol, ethynylphenylcarbinol, phenylethynylcarbinol, 2-propyn-1-ol, 1-phenyl, benzyl alcohol, .alpha.-ethynyl, benzenemethanol, .alpha.-ethynyl, alpha-ethynylbenzyl alcohol, alpha-ethynylbenzenemethanol, alpha-phenylpropargyl alcohol
1-phenylprop-2-yn-1-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 224-064-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only