missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-2-propyn-1-ol, 98%

Catalog No. AAL0954906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0954906 5 g
AAL0954903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0954906 Supplier Thermo Scientific Chemicals Supplier No. L0954906

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4187-87-5 | C9H8O | 132.162 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4187-87-5
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.162
MDL Number MFCD00021860
InChI Key UIGLAZDLBZDVBL-UHFFFAOYSA-N
Synonym 1-phenyl-2-propyn-1-ol, 1-phenylpropargyl alcohol, ethynylphenylcarbinol, phenylethynylcarbinol, 2-propyn-1-ol, 1-phenyl, benzyl alcohol, .alpha.-ethynyl, benzenemethanol, .alpha.-ethynyl, alpha-ethynylbenzyl alcohol, alpha-ethynylbenzenemethanol, alpha-phenylpropargyl alcohol
PubChem CID 20155
IUPAC Name 1-phenylprop-2-yn-1-ol
SMILES C#CC(C1=CC=CC=C1)O

Specifications

Melting Point 26°C to 30°C
Density 1.087
Boiling Point 231°C to 233°C
Flash Point 99°C (210°F)
Refractive Index 1.55
Quantity 5 g
Beilstein 742365
Formula Weight 132.16
Percent Purity 98%
Chemical Name or Material 1-Phenyl-2-propyn-1-ol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 224-064-6
RTECSNumber DO5900000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only