missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-2-butanone 95.0+%, TCI America™

Catalog No. P09715ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P09715ML 5 mL
P097125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P09715ML Supplier TCI America Supplier No. P09715ML
Only null left

Chemical Identifiers

CAS 1007-32-5
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD00009315
InChI Key GKDLTXYXODKDEA-UHFFFAOYSA-N
Synonym 1-phenyl-2-butanone, benzyl ethyl ketone, 2-butanone, 1-phenyl, unii-1akg6904vp, ethyl benzyl ketone, 2-butanone, phenyl, phenylbutanone, banzylethylketone, 1-phenyl-butanone, 2-oxo-1-phenylbutane
PubChem CID 13879
IUPAC Name 1-phenylbutan-2-one
SMILES CCC(=O)CC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 226°C
Quantity 5 mL
Formula Weight 148.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Phenyl-2-butanone