Learn More
1-Phenyl-1H-pyrazole, 98%
CAS: 1126-00-7 | C9H8N2 | 144.18 g/mol
Supplier: Thermo Scientific Chemicals A1427030
Description
1-Phenyl-1H-pyrazole is used in the preparation of 4,5-diphenylpyrazolo[1,5-a]quinoline, 1-(1,2,3,4-tetraphenylnaphthalen-5-yl)pyrazole and 1-(1,2,3,4,5,6,7,8-octaphenylanthracen-9-yl)pyrazole. It acts as a cyclometallated ligand in the preparation of new heteroleptic iridium(III) complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Phenyl-1H-pyrazole is used in the preparation of 4,5-diphenylpyrazolo[1,5-a]quinoline, 1-(1,2,3,4-tetraphenylnaphthalen-5-yl)pyrazole and 1-(1,2,3,4,5,6,7,8-octaphenylanthracen-9-yl)pyrazole. It acts as a cyclometallated ligand in the preparation of new heteroleptic iridium(III) complexes.
Solubility
Soluble in water.
Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Specifications
| 1-Phenyl-1H-pyrazole | |
| 1.09 | |
| 109°C (228°F) | |
| 1.596 | |
| 250 g | |
| 1-phenyl-1h-pyrazole, n-phenylpyrazole, 1h-pyrazole, 1-phenyl, pyrazole, 1-phenyl, 1-phenylpyrazol, n-phenyl pyrazole, 1-phenyl pyrazole, 1-phenyl-pyrazole, 1-phenyl-1h-pyrazol, l-phenyl-1h-pyrazole | |
| WITMXBRCQWOZPX-UHFFFAOYSA-N | |
| 1-phenylpyrazole | |
| 70769 | |
| 144.18 |
| 1126-00-7 | |
| 141°C to 142°C (30 mmHg) | |
| C9H8N2 | |
| MFCD00003112 | |
| 2594 | |
| Soluble in water. | |
| C1=CN(N=C1)C1=CC=CC=C1 | |
| 144.18 | |
| CHEBI:38879 | |
| 98% |
Chemical Identifiers
| 1126-00-7 | |
| 144.18 | |
| WITMXBRCQWOZPX-UHFFFAOYSA-N | |
| 70769 | |
| 1-phenylpyrazole |
| C9H8N2 | |
| MFCD00003112 | |
| 1-phenyl-1h-pyrazole, n-phenylpyrazole, 1h-pyrazole, 1-phenyl, pyrazole, 1-phenyl, 1-phenylpyrazol, n-phenyl pyrazole, 1-phenyl pyrazole, 1-phenyl-pyrazole, 1-phenyl-1h-pyrazol, l-phenyl-1h-pyrazole | |
| CHEBI:38879 | |
| C1=CN(N=C1)C1=CC=CC=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 214-415-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only