missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1H-pyrazole, 98%

Catalog No. AAA1427009
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1427009 10 g
AAA1427018 50 g
AAA1427030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1427009 Supplier Thermo Scientific Chemicals Supplier No. A1427009
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1126-00-7 | C9H8N2 | 144.18 g/mol

1-Phenyl-1H-pyrazole is used in the preparation of 4,5-diphenylpyrazolo[1,5-a]quinoline, 1-(1,2,3,4-tetraphenylnaphthalen-5-yl)pyrazole and 1-(1,2,3,4,5,6,7,8-octaphenylanthracen-9-yl)pyrazole. It acts as a cyclometallated ligand in the preparation of new heteroleptic iridium(III) complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Phenyl-1H-pyrazole is used in the preparation of 4,5-diphenylpyrazolo[1,5-a]quinoline, 1-(1,2,3,4-tetraphenylnaphthalen-5-yl)pyrazole and 1-(1,2,3,4,5,6,7,8-octaphenylanthracen-9-yl)pyrazole. It acts as a cyclometallated ligand in the preparation of new heteroleptic iridium(III) complexes.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.

Chemical Identifiers

CAS 1126-00-7
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.18
MDL Number MFCD00003112
InChI Key WITMXBRCQWOZPX-UHFFFAOYSA-N
Synonym 1-phenyl-1h-pyrazole, n-phenylpyrazole, 1h-pyrazole, 1-phenyl, pyrazole, 1-phenyl, 1-phenylpyrazol, n-phenyl pyrazole, 1-phenyl pyrazole, 1-phenyl-pyrazole, 1-phenyl-1h-pyrazol, l-phenyl-1h-pyrazole
PubChem CID 70769
ChEBI CHEBI:38879
IUPAC Name 1-phenylpyrazole
SMILES C1=CN(N=C1)C1=CC=CC=C1

Specifications

Density 1.09
Boiling Point 141°C to 142°C (30 mmHg)
Flash Point 109°C (228°F)
Refractive Index 1.596
Quantity 10 g
Beilstein 2594
Solubility Information Soluble in water.
Formula Weight 144.18
Percent Purity 98%
Chemical Name or Material 1-Phenyl-1H-pyrazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-415-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only