missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Phenyl-1H-pyrazole-5-carbaldehyde, 95+%, Thermo Scientific™
Chemical Identifiers
| CAS | 132274-70-5 |
|---|---|
| Molecular Formula | C10H8N2O |
| Molecular Weight (g/mol) | 172.187 |
| InChI Key | OQORFMABOZEDBL-UHFFFAOYSA-N |
| Synonym | 1-phenyl-1h-pyrazole-5-carbaldehyde, 1h-pyrazole-5-carboxaldehyde, 1-phenyl, 1-phenylpyrazole-5-carbaldehyde, 1-phenyl-1h-pyrazole-5-carboxaldehyde |
| PubChem CID | 2776567 |
| IUPAC Name | 2-phenylpyrazole-3-carbaldehyde |
| SMILES | C1=CC=C(C=C1)N2C(=CC=N2)C=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
CC24504CB
|
N/A | 250mg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC24504DA
|
N/A | 1g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC24504DE
|
N/A | 5g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 132274-70-5 | |
| 172.187 | |
| 1-phenyl-1h-pyrazole-5-carbaldehyde, 1h-pyrazole-5-carboxaldehyde, 1-phenyl, 1-phenylpyrazole-5-carbaldehyde, 1-phenyl-1h-pyrazole-5-carboxaldehyde | |
| 2-phenylpyrazole-3-carbaldehyde |
| C10H8N2O | |
| OQORFMABOZEDBL-UHFFFAOYSA-N | |
| 2776567 | |
| C1=CC=C(C=C1)N2C(=CC=N2)C=O |
Specifications
| 132274-70-5 | |
| C10H8N2O | |
| OQORFMABOZEDBL-UHFFFAOYSA-N | |
| 2-phenylpyrazole-3-carbaldehyde | |
| 2776567 | |
| 95% |
| 95% | |
| 1-phenyl-1h-pyrazole-5-carbaldehyde, 1h-pyrazole-5-carboxaldehyde, 1-phenyl, 1-phenylpyrazole-5-carbaldehyde, 1-phenyl-1h-pyrazole-5-carboxaldehyde | |
| C1=CC=C(C=C1)N2C(=CC=N2)C=O | |
| 172.187 | |
| 172.19 | |
| 1-Phenyl-1H-pyrazole-5-carbaldehyde |