Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Phenyl-1H-pyrazole-5-carbaldehyde, 95+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC24504DE
| Quantity | 5g |
|---|---|
| Packaging | Amber Glass Bottle |
Chemical Identifiers
| 132274-70-5 | |
| 172.187 | |
| 1-phenyl-1h-pyrazole-5-carbaldehyde, 1h-pyrazole-5-carboxaldehyde, 1-phenyl, 1-phenylpyrazole-5-carbaldehyde, 1-phenyl-1h-pyrazole-5-carboxaldehyde | |
| 2-phenylpyrazole-3-carbaldehyde |
| C10H8N2O | |
| OQORFMABOZEDBL-UHFFFAOYSA-N | |
| 2776567 | |
| C1=CC=C(C=C1)N2C(=CC=N2)C=O |
Specifications
| 1-Phenyl-1H-pyrazole-5-carbaldehyde | |
| 95% | |
| C10H8N2O | |
| 1-phenyl-1h-pyrazole-5-carbaldehyde, 1h-pyrazole-5-carboxaldehyde, 1-phenyl, 1-phenylpyrazole-5-carbaldehyde, 1-phenyl-1h-pyrazole-5-carboxaldehyde | |
| C1=CC=C(C=C1)N2C(=CC=N2)C=O | |
| 172.187 | |
| 172.19 |
| 132274-70-5 | |
| Amber Glass Bottle | |
| 5g | |
| OQORFMABOZEDBL-UHFFFAOYSA-N | |
| 2-phenylpyrazole-3-carbaldehyde | |
| 2776567 | |
| 95% |