missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-trimethylsiloxyethylene, 95%

Catalog No. AAL0530803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0530803 1 g
AAL0530806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0530803 Supplier Thermo Scientific Chemicals Supplier No. L0530803
Only null left

CAS: 13735-81-4 | C11H16OSi | 192.333 g/mol

1-Phenyl-1-trimethylsiloxyethylene has been used in the synthesis of β-amino ketones in water via Mannich-type reaction and cis-2,6-disubstituted dihydropyrans via three-component, one-pot cascade reaction. It is also used as chemical additives and intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Phenyl-1-trimethylsiloxyethylene has been used in the synthesis of β-amino ketones in water via Mannich-type reaction and cis-2,6-disubstituted dihydropyrans via three-component, one-pot cascade reaction. It is also used as chemical additives and intermediates.

Solubility
Reacts with water.

Notes
Store at 4°C. Moisture Sensitive. Store under inert gas. Incompatible materials are Oxidizing agents.

Chemical Identifiers

CAS 13735-81-4
Molecular Formula C11H16OSi
Molecular Weight (g/mol) 192.333
MDL Number MFCD00008582
InChI Key AFFPCIMDERUIST-UHFFFAOYSA-N
Synonym 1-phenyl-1-trimethylsiloxyethylene, trimethyl 1-phenylvinyl oxy silane, 1-phenyl-1-trimethylsilyloxyethylene, 1-phenyl-1-trimethylsiloxy ethylene, 1-phenyl-1-trimethylsilyloxy ethylene, benzene, 1-trimethylsilyl oxy ethenyl, silane, trimethyl 1-phenylethenyl oxy, acmc-209ccf
PubChem CID 117406
IUPAC Name trimethyl(1-phenylethenoxy)silane
SMILES C[Si](C)(C)OC(=C)C1=CC=CC=C1

Specifications

Boiling Point 53°C to 54°C (1mmHg)
Quantity 1 g
Beilstein 1306914
Flash Point 78°C (172°F)
Solubility Information Reacts with water.
Formula Weight 192.34
Percent Purity 97%
Refractive Index 1.502
Sensitivity Moisture sensitive
Density 0.938
Chemical Name or Material 1-Phenyl-1-trimethylsiloxyethylene
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 237-308-1
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only