Learn More
1-Phenyl-1-pyridin-2-ylmethanamine dihydrochloride, 95%, Thermo Scientific™
$406.25 - $406.25
Chemical Identifiers
| CAS | 59575-91-6 |
|---|---|
| Molecular Formula | C12H13ClN2 |
| Molecular Weight (g/mol) | 220.70 |
| MDL Number | MFCD00102147 |
| InChI Key | GVGSFONXPJCBIS-UHFFFAOYNA-N |
| Synonym | phenyl pyridin-2-yl methanamine hydrochloride, 1-phenyl-1-pyridin-2-ylmethanamine hydrochloride, phenyl 2-pyridyl methylamine hydrochloride, phenyl-2-pyridylmethylamine, chloride, phenyl 2-pyridyl methanamine hydrochloride, phenyl pyridin-2-yl methylamine hydrochloride, alpha-pyridin-2-yl benzylamine hydrochloride, phenyl 2-pyridyl methylamine dihydrochloride, 1-phenyl-1-pyridin-2-yl methanamine hydrochloride |
| PubChem CID | 2775257 |
| IUPAC Name | phenyl(pyridin-2-yl)methanamine;hydrochloride |
| SMILES | Cl.NC(C1=CC=CC=C1)C1=CC=CC=N1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
BTB10358DA
|
Thermo Scientific Chemicals
BTB10358DA |
1 g | Amber Glass Bottle |
Each for $406.25
|
|
||||
Chemical Identifiers
| 59575-91-6 | |
| 220.70 | |
| GVGSFONXPJCBIS-UHFFFAOYNA-N | |
| 2775257 | |
| Cl.NC(C1=CC=CC=C1)C1=CC=CC=N1 |
| C12H13ClN2 | |
| MFCD00102147 | |
| phenyl pyridin-2-yl methanamine hydrochloride, 1-phenyl-1-pyridin-2-ylmethanamine hydrochloride, phenyl 2-pyridyl methylamine hydrochloride, phenyl-2-pyridylmethylamine, chloride, phenyl 2-pyridyl methanamine hydrochloride, phenyl pyridin-2-yl methylamine hydrochloride, alpha-pyridin-2-yl benzylamine hydrochloride, phenyl 2-pyridyl methylamine dihydrochloride, 1-phenyl-1-pyridin-2-yl methanamine hydrochloride | |
| phenyl(pyridin-2-yl)methanamine;hydrochloride |
Specifications
| 59575-91-6 | |
| 100.0 | |
| Amber Glass Bottle | |
| MFCD00102147 | |
| phenyl pyridin-2-yl methanamine hydrochloride, 1-phenyl-1-pyridin-2-ylmethanamine hydrochloride, phenyl 2-pyridyl methylamine hydrochloride, phenyl-2-pyridylmethylamine, chloride, phenyl 2-pyridyl methanamine hydrochloride, phenyl pyridin-2-yl methylamine hydrochloride, alpha-pyridin-2-yl benzylamine hydrochloride, phenyl 2-pyridyl methylamine dihydrochloride, 1-phenyl-1-pyridin-2-yl methanamine hydrochloride | |
| Cl.NC(C1=CC=CC=C1)C1=CC=CC=N1 | |
| 220.70 | |
| 257.16 | |
| 1-Phenyl-1-pyridin-2-ylmethanamine dihydrochloride |
| 94.5 | |
| 95% | |
| C12H13ClN2 | |
| 1 g | |
| GVGSFONXPJCBIS-UHFFFAOYNA-N | |
| phenyl(pyridin-2-yl)methanamine;hydrochloride | |
| 2775257 | |
| 95% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning