missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-pyridin-2-ylmethanamine dihydrochloride, 95%, Thermo Scientific™

Catalog No. BTB10358DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BTB10358DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTB10358DA Supplier Thermo Scientific Chemicals Supplier No. BTB10358DA

Chemical Identifiers

CAS 59575-91-6
Molecular Formula C12H13ClN2
Molecular Weight (g/mol) 220.70
MDL Number MFCD00102147
InChI Key GVGSFONXPJCBIS-UHFFFAOYNA-N
Synonym phenyl pyridin-2-yl methanamine hydrochloride, 1-phenyl-1-pyridin-2-ylmethanamine hydrochloride, phenyl 2-pyridyl methylamine hydrochloride, phenyl-2-pyridylmethylamine, chloride, phenyl 2-pyridyl methanamine hydrochloride, phenyl pyridin-2-yl methylamine hydrochloride, alpha-pyridin-2-yl benzylamine hydrochloride, phenyl 2-pyridyl methylamine dihydrochloride, 1-phenyl-1-pyridin-2-yl methanamine hydrochloride
PubChem CID 2775257
IUPAC Name phenyl(pyridin-2-yl)methanamine;hydrochloride
SMILES Cl.NC(C1=CC=CC=C1)C1=CC=CC=N1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 257.16
Percent Purity 95%
Chemical Name or Material 1-Phenyl-1-pyridin-2-ylmethanamine dihydrochloride
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.