missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-pentyne, 98%

Catalog No. p-7025405
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2085706 5 g
AAB2085714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2085706 Supplier Thermo Scientific Chemicals Supplier No. B2085706

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4250-81-1 | C11H12 | 144.22 g/mol

It finds it application in the highly regioselective synthesis of polysubstituted naphthalene derivatives through gallium trichloride catalyzed alkyne-aldehyde coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds it application in the highly regioselective synthesis of polysubstituted naphthalene derivatives through gallium trichloride catalyzed alkyne-aldehyde coupling.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 4250-81-1
Molecular Formula C11H12
Molecular Weight (g/mol) 144.22
MDL Number MFCD00039959
InChI Key DEGIOKWPYFOHGH-UHFFFAOYSA-N
Synonym 1-phenyl-1-pentyne, pent-1-yn-1-ylbenzene, 1-pentynylbenzene, benzene, 1-pentynyl, 1-pentynylbenzene #, acmc-1aso7, 1-phenyl-2-propylacetylene
PubChem CID 77923
IUPAC Name pent-1-ynylbenzene
SMILES CCCC#CC1=CC=CC=C1

Specifications

Density 0.906
Boiling Point 213°C to 215°C
Flash Point 82°C (179°F)
Refractive Index 1.5413
Linear Formula CH3CH2CH2C≡CC6H5
Quantity 5 g
Beilstein 1853156
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 144.22
Percent Purity 98%
Chemical Name or Material 1-Phenyl-1-pentyne
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 224-211-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only