missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-pentyn-3-ol, 97%

Catalog No. p-7035692
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5341603 1 g
AAH5341606 5 g
AAH5341614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5341603 Supplier Thermo Scientific Chemicals Supplier No. H5341603
Only null left

CAS: 27975-78-6 | C11H12O | 160.216 g/mol

Reactant in the heterogeneously catalyzed etherification of glycerol, also in fritsch-wiechell rearrangement of 1-chlorovinyl sulfoxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant in the heterogeneously catalyzed etherification of glycerol, also in fritsch-wiechell rearrangement of 1-chlorovinyl sulfoxides.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 27975-78-6
Molecular Formula C11H12O
Molecular Weight (g/mol) 160.216
MDL Number MFCD00041589
InChI Key QWCMSASONHVIHV-UHFFFAOYSA-N
Synonym 1-phenyl-1-pentyn-3-ol, 1-pentyn-3-ol,1-phenyl, 1-pentyn-3-ol, 1-phenyl-, s, acmc-20mqra, 1-pentyn-3-ol, 1-phenyl
PubChem CID 11789508
IUPAC Name 1-phenylpent-1-yn-3-ol
SMILES CCC(C#CC1=CC=CC=C1)O

Specifications

Density 1.336
Flash Point >110°C (230°F)
Refractive Index 1.559
Quantity 1 g
Formula Weight 160.22
Percent Purity 97%
Chemical Name or Material 1-Phenyl-1-pentyn-3-ol
Hazard Category H302-H319
Hazard Statement GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only