missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-hexyne 98.0+%, TCI America™

Catalog No. P03605ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P03605ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P03605ML Supplier TCI America Supplier No. P03605ML

Chemical Identifiers

CAS 1129-65-3
Molecular Formula C12H14
Molecular Weight (g/mol) 158.244
MDL Number MFCD00039969
InChI Key VBRLZTLFLNZEPZ-UHFFFAOYSA-N
Synonym 1-Butyl-2-phenylacetylene
PubChem CID 136908
IUPAC Name hex-1-ynylbenzene
SMILES CCCCC#CC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 231°C
Quantity 5 mL
Formula Weight 158.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Phenyl-1-hexyne