missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Phenyl-1-cyclopropanecarboxylic Acid 98.0+%, TCI America™
$177.08 - $177.08
Chemical Identifiers
| CAS | 6120-95-2 |
|---|---|
| Molecular Formula | C10H10O2 |
| Molecular Weight (g/mol) | 162.188 |
| MDL Number | MFCD00001288 |
| InChI Key | IWWCCNVRNHTGLV-UHFFFAOYSA-N |
| Synonym | 1-phenylcyclopropanecarboxylic acid, 1-phenyl-1-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-phenyl, 1-phenyl-cyclopropanecarboxylic acid, 1-phenyl-1-cyclopropanecarboxylicacid, pubchem15554, acmc-1b2kn, phenylcyclopropanecarboxylic acid, 1-phenyl-cyclopropanecarboxlic acid, 1-phenylcyclo-propanecarboxylic acid |
| PubChem CID | 80206 |
| IUPAC Name | 1-phenylcyclopropane-1-carboxylic acid |
| SMILES | C1CC1(C2=CC=CC=C2)C(=O)O |
Chemical Identifiers
| 6120-95-2 | |
| 162.188 | |
| IWWCCNVRNHTGLV-UHFFFAOYSA-N | |
| 80206 | |
| C1CC1(C2=CC=CC=C2)C(=O)O |
| C10H10O2 | |
| MFCD00001288 | |
| 1-phenylcyclopropanecarboxylic acid, 1-phenyl-1-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-phenyl, 1-phenyl-cyclopropanecarboxylic acid, 1-phenyl-1-cyclopropanecarboxylicacid, pubchem15554, acmc-1b2kn, phenylcyclopropanecarboxylic acid, 1-phenyl-cyclopropanecarboxlic acid, 1-phenylcyclo-propanecarboxylic acid | |
| 1-phenylcyclopropane-1-carboxylic acid |
Specifications
| 6120-95-2 | |
| White-Yellow | |
| MFCD00001288 | |
| 1-phenylcyclopropanecarboxylic acid, 1-phenyl-1-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-phenyl, 1-phenyl-cyclopropanecarboxylic acid, 1-phenyl-1-cyclopropanecarboxylicacid, pubchem15554, acmc-1b2kn, phenylcyclopropanecarboxylic acid, 1-phenyl-cyclopropanecarboxlic acid, 1-phenylcyclo-propanecarboxylic acid | |
| C1CC1(C2=CC=CC=C2)C(=O)O | |
| 162.188 | |
| 162.19 | |
| Crystalline Powder |
| 86°C | |
| C10H10O2 | |
| 5 g | |
| IWWCCNVRNHTGLV-UHFFFAOYSA-N | |
| 1-phenylcyclopropane-1-carboxylic acid | |
| 80206 | |
| ≥98.0% (GC,T) | |
| 1-Phenyl-1-cyclopropanecarboxylic Acid |
Safety and Handling
TSCA : No