missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Phenyl-1-cyclopropanecarboxylic acid, 97%
CAS: 6120-95-2 | C10H10O2 | 162.19 g/mol
$155.87 - $155.87
Chemical Identifiers
| CAS | 6120-95-2 |
|---|---|
| Molecular Formula | C10H10O2 |
| Molecular Weight (g/mol) | 162.19 |
| MDL Number | MFCD00001288 |
| InChI Key | IWWCCNVRNHTGLV-UHFFFAOYSA-N |
| Synonym | 1-phenylcyclopropanecarboxylic acid, 1-phenyl-1-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-phenyl, 1-phenyl-cyclopropanecarboxylic acid, 1-phenyl-1-cyclopropanecarboxylicacid, pubchem15554, acmc-1b2kn, phenylcyclopropanecarboxylic acid, 1-phenyl-cyclopropanecarboxlic acid, 1-phenylcyclo-propanecarboxylic acid |
| PubChem CID | 80206 |
| IUPAC Name | 1-phenylcyclopropane-1-carboxylic acid |
| SMILES | C1CC1(C2=CC=CC=C2)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC170680100
|
Thermo Scientific Chemicals
170680100 |
10 g | Glass bottle |
Each for $155.87
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 6120-95-2 | |
| 162.19 | |
| IWWCCNVRNHTGLV-UHFFFAOYSA-N | |
| 80206 | |
| C1CC1(C2=CC=CC=C2)C(=O)O |
| C10H10O2 | |
| MFCD00001288 | |
| 1-phenylcyclopropanecarboxylic acid, 1-phenyl-1-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-phenyl, 1-phenyl-cyclopropanecarboxylic acid, 1-phenyl-1-cyclopropanecarboxylicacid, pubchem15554, acmc-1b2kn, phenylcyclopropanecarboxylic acid, 1-phenyl-cyclopropanecarboxlic acid, 1-phenylcyclo-propanecarboxylic acid | |
| 1-phenylcyclopropane-1-carboxylic acid |
Specifications
| 6120-95-2 | |
| 100.0 | |
| White to Beige | |
| 96% min. (GC) | |
| C10H10O2 | |
| MFCD00001288 | |
| 1-phenylcyclopropanecarboxylic acid, 1-phenyl-1-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-phenyl, 1-phenyl-cyclopropanecarboxylic acid, 1-phenyl-1-cyclopropanecarboxylicacid, pubchem15554, acmc-1b2kn, phenylcyclopropanecarboxylic acid, 1-phenyl-cyclopropanecarboxlic acid, 1-phenylcyclo-propanecarboxylic acid | |
| C1CC1(C2=CC=CC=C2)C(=O)O | |
| 162.19 | |
| 162.19 | |
| Crystalline Powder, Crystals or Chunks |
| 96.0 | |
| 84°C to 87°C | |
| Authentic | |
| Glass bottle | |
| C6H5C3H4CO2H | |
| 10 g | |
| IWWCCNVRNHTGLV-UHFFFAOYSA-N | |
| 1-phenylcyclopropane-1-carboxylic acid | |
| 80206 | |
| 97% | |
| 1-Phenyl-1-cyclopropanecarboxylic acid, 97% |
Safety and Handling
EINECSNumber : 228-090-9
TSCA : TSCA
RUO – Research Use Only