missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-cyclopropanecarbonitrile, 97%

Catalog No. AC170590100
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC170590100 10 g Glass bottle
AC170590500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC170590100 Supplier Thermo Scientific Chemicals Supplier No. 170590100

CAS: 935-44-4 | C10H9N | 143.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 935-44-4
Molecular Formula C10H9N
Molecular Weight (g/mol) 143.19
MDL Number MFCD00001270
InChI Key ZHFURHRJUWYDKG-UHFFFAOYSA-N
Synonym 1-phenylcyclopropanecarbonitrile, 1-phenyl-1-cyclopropanecarbonitrile, cyclopropanecarbonitrile, 1-phenyl, 1-cyano-1-phenylcyclopropane, 1-phenyl-cyclopropanecarbonitrile, 1-phenylcyclopropyl cyanide, phenylcyclopropanenitrile, 1-phenylcyclopropanenitrile, phenylcyclopropanecarbonitrile, 1-phenylcyclopropane carbonitrile
PubChem CID 70288
IUPAC Name 1-phenylcyclopropane-1-carbonitrile
SMILES N#CC1(CC1)C1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0000g/mL
Boiling Point 133.0°C to 137.0°C (30.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5370 to 1.5400
Linear Formula C6H5C3H4CN
Quantity 10 g
Specific Gravity 1
Formula Weight 143.19
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Phenyl-1-cyclopropanecarbonitrile, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 213-304-5
TSCA TSCA

RUO – Research Use Only