missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Phenyl-1-cyclopentanecarboxylic Acid 98.0+%, TCI America™
$48.46 - $141.67
Chemical Identifiers
| CAS | 77-55-4 |
|---|---|
| Molecular Formula | C12H14O2 |
| Molecular Weight (g/mol) | 190.24 |
| MDL Number | MFCD00001372 |
| InChI Key | RHPCYZLXNNRRMB-UHFFFAOYSA-N |
| Synonym | 1-phenylcyclopentanecarboxylic acid, 1-phenyl-1-cyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 1-phenyl, 1-phenyl-cyclopentanecarboxylic acid, 1-phenylcyclopentane carboxylic acid, cyclopentanecarboxylicacid, 1-phenyl, pubchem22923, acmc-209pao, phenylcyclopentanecarboxylic acid, phenylcyclopentane carboxylic acid |
| PubChem CID | 66167 |
| IUPAC Name | 1-phenylcyclopentane-1-carboxylic acid |
| SMILES | OC(=O)C1(CCCC1)C1=CC=CC=C1 |
Chemical Identifiers
| 77-55-4 | |
| 190.24 | |
| RHPCYZLXNNRRMB-UHFFFAOYSA-N | |
| 66167 | |
| OC(=O)C1(CCCC1)C1=CC=CC=C1 |
| C12H14O2 | |
| MFCD00001372 | |
| 1-phenylcyclopentanecarboxylic acid, 1-phenyl-1-cyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 1-phenyl, 1-phenyl-cyclopentanecarboxylic acid, 1-phenylcyclopentane carboxylic acid, cyclopentanecarboxylicacid, 1-phenyl, pubchem22923, acmc-209pao, phenylcyclopentanecarboxylic acid, phenylcyclopentane carboxylic acid | |
| 1-phenylcyclopentane-1-carboxylic acid |
Specifications
| 77-55-4 | |
| White-Yellow | |
| MFCD00001372 | |
| 1-phenylcyclopentanecarboxylic acid, 1-phenyl-1-cyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 1-phenyl, 1-phenyl-cyclopentanecarboxylic acid, 1-phenylcyclopentane carboxylic acid, cyclopentanecarboxylicacid, 1-phenyl, pubchem22923, acmc-209pao, phenylcyclopentanecarboxylic acid, phenylcyclopentane carboxylic acid | |
| OC(=O)C1(CCCC1)C1=CC=CC=C1 | |
| 190.24 | |
| 190.24 | |
| Crystalline Powder |
| 159°C | |
| C12H14O2 | |
| 5 g | |
| RHPCYZLXNNRRMB-UHFFFAOYSA-N | |
| 1-phenylcyclopentane-1-carboxylic acid | |
| 66167 | |
| ≥98.0% (GC,T) | |
| 1-Phenyl-1-cyclopentanecarboxylic Acid |
Safety and Handling
EINECSNumber : (9)-1186
TSCA : No