missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-cyclohexene 95.0+%, TCI America™

Catalog No. p-7135065
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13025ML 5 mL
P130225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13025ML Supplier TCI America Supplier No. P13025ML
Only null left

Chemical Identifiers

CAS 771-98-2
Molecular Formula C12H14
Molecular Weight (g/mol) 158.244
MDL Number MFCD00001542
InChI Key WCMSFBRREKZZFL-UHFFFAOYSA-N
Synonym 1-phenyl-1-cyclohexene, 1-phenylcyclohexene, 1-phenylcyclohex-1-ene, benzene, 1-cyclohexen-1-yl, phenylcyclohexene, cyclohexenylbenzene, benzene, cyclohexenyl, 2,3,4,5-tetrahydro-1,1'-biphenyl, cyclohexene, 1-phenyl, trans-1-phenylcyclohexene
PubChem CID 13043
IUPAC Name cyclohexen-1-ylbenzene
SMILES C1CCC(=CC1)C2=CC=CC=C2

Specifications

Color Colorless
Boiling Point 252°C
Quantity 5 mL
Formula Weight 158.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Phenyl-1-cyclohexene
RTECSNumber GW6600000
TSCA No