missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Phenyl-1-butanol 98.0+%, TCI America™
Supplier: TCI America P168225G
Specifications
| 1-Phenyl-1-butanol | |
| 16°C | |
| 170°C | |
| MFCD00066173 | |
| alpha-Propylbenzyl Alcohol | |
| CCCC(C1=CC=CC=C1)O | |
| 150.221 | |
| 150.22 | |
| Liquid |
| 614-14-2 | |
| Yellow | |
| C10H14O | |
| 25 g | |
| HQRWWHIETAKIMO-UHFFFAOYSA-N | |
| 1-phenylbutan-1-ol | |
| 95372 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 614-14-2 | |
| 150.221 | |
| HQRWWHIETAKIMO-UHFFFAOYSA-N | |
| 95372 | |
| CCCC(C1=CC=CC=C1)O |
| C10H14O | |
| MFCD00066173 | |
| alpha-Propylbenzyl Alcohol | |
| 1-phenylbutan-1-ol |
Safety and Handling
EINECSNumber : (3)-1034
TSCA : No