missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1,3-butanedione 98.0+%, TCI America™

Catalog No. P0160500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0160500G 500 g
P016025G 25 g
P0160100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P0160500G Supplier TCI America Supplier No. P0160500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 93-91-4
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00008786
InChI Key CVBUKMMMRLOKQR-UHFFFAOYSA-N
Synonym benzoylacetone, 1-phenyl-1,3-butanedione, 1-benzoylacetone, 1,3-butanedione, 1-phenyl, 1-benzoyl-2-propanone, acetoacetophenone, 2-propanone, benzoyl, acetylbenzoylmethane, 2-acetylacetophenone, benzoyl-aceton
PubChem CID 7166
IUPAC Name 1-phenylbutane-1,3-dione
SMILES CC(=O)CC(=O)C1=CC=CC=C1

Specifications

Melting Point 57°C
Color White-Yellow
Boiling Point 262°C
Quantity 500 g
Formula Weight 162.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Phenyl-1,3-butanedione
EINECSNumber (3)-3494
RTECSNumber EK3540200
TSCA Yes