missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-1-Phenyl-1,2-ethanediol, 97%

Catalog No. AAL0455106
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0455106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0455106 Supplier Thermo Scientific Chemicals Supplier No. L0455106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 93-56-1 | C8H10O2 | 138.166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 93-56-1
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00003546
InChI Key PWMWNFMRSKOCEY-UHFFFAOYSA-N
Synonym 1-phenyl-1,2-ethanediol, styrene glycol, phenylethylene glycol, phenylethanediol, phenyl glycol, 1,2-ethanediol, 1-phenyl, phenyl-1,2-ethanediol, styrolyl alcohol, fenylglycol, 1-phenylethylene glycol
PubChem CID 7149
IUPAC Name 1-phenylethane-1,2-diol
SMILES C1=CC=C(C=C1)C(CO)O

Specifications

Melting Point 66°C to 68°C
Boiling Point 272°C to 274°C
Flash Point 160°C (320°F)
Quantity 5 g
Beilstein 1306723
Merck Index 14,8861
Formula Weight 138.17
Percent Purity 97%
Chemical Name or Material (+/-)-1-Phenyl-1,2-ethanediol
EINECSNumber 202-258-1
RTECSNumber KI2500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only