missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| 1-Pentyne | |
| Yellow | |
| C5H8 | |
| 25 mL | |
| 1-pentyne, propylacetylene, pentyne, acetylene, propyl, pentin, pentyn, 1-n-pentyne, pent-1-yn-1-yl, acmc-1cuk3 | |
| CCCC#C | |
| 68.12 | |
| 68.12 | |
| Liquid |
| 627-19-0 | |
| 40°C | |
| MFCD00009469 | |
| 3295 | |
| IBXNCJKFFQIKKY-UHFFFAOYSA-N | |
| pent-1-yne | |
| 12309 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 627-19-0 | |
| 68.12 | |
| IBXNCJKFFQIKKY-UHFFFAOYSA-N | |
| 12309 | |
| CCCC#C |
| C5H8 | |
| MFCD00009469 | |
| 1-pentyne, propylacetylene, pentyne, acetylene, propyl, pentin, pentyn, 1-n-pentyne, pent-1-yn-1-yl, acmc-1cuk3 | |
| pent-1-yne |
Safety and Handling
TSCA : Yes