missing translation for 'onlineSavingsMsg'
Learn More

1-Pentylamine, 98%

CAS: 110-58-7 | C5H13N | 87.166 g/mol

Supplier:  Thermo Scientific Chemicals A1167422

 View more versions of this product

Catalog No. AAA1167422


Only null left
Explore available promotions

Description

Description

1-Pentylamine is used as a solvent, as a raw material in the manufacture of a variety of other compounds, including dyes, emulsifiers, and pharmaceutical products and as a flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pentylamine is used as a solvent, as a raw material in the manufacture of a variety of other compounds, including dyes, emulsifiers, and pharmaceutical products and as a flavoring agent.

Solubility
Soluble in alcohol, ether.

Notes
Air Sensitive. Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

1-Pentylamine
-55°C
103°C to 105°C
Amine-like
1.411
100 g
505953
14,598
Soluble in alcohol,ether.
CCCCCN
87.166
CHEBI:74848
98%
110-58-7
0.751
−1°C (30°F)
C5H13N
MFCD00008236
UN1106
Air Sensitive
amylamine, 1-aminopentane, pentylamine, 1-pentylamine, n-amylamine, 1-pentanamine, n-pentylamine, monoamylamine, norleucamine, amyl amine
DPBLXKKOBLCELK-UHFFFAOYSA-N
pentan-1-amine
8060
87.17

Chemical Identifiers

110-58-7
87.166
DPBLXKKOBLCELK-UHFFFAOYSA-N
8060
pentan-1-amine
C5H13N
MFCD00008236
amylamine, 1-aminopentane, pentylamine, 1-pentylamine, n-amylamine, 1-pentanamine, n-pentylamine, monoamylamine, norleucamine, amyl amine
CHEBI:74848
CCCCCN
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H331-H314-H318-H302-H312
Highly flammable liquid and vapor.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H302+H312-H314-H331-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMYLAMINES

EINECSNumber : 203-780-2

RTECSNumber : SC0300000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only