missing translation for 'onlineSavingsMsg'
Learn More

1-Penten-3-yl Acetate 97.0+%, TCI America™

Supplier:  TCI America A15425ML

Catalog No. A15425ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1-Penten-3-yl Acetate
Yellow
C7H12O2
5 mL
Acetic Acid 1-Ethylallyl Ester, Acetic Acid 1-Penten-3-yl Ester, 1-Ethylallyl Acetate
CCC(C=C)OC(=O)C
128.171
128.17
Liquid
10500-11-5
132°C
MFCD02093427
3272
MRLKTTBPWZXARX-UHFFFAOYSA-N
pent-1-en-3-yl acetate
530323
≥97.0% (GC)

Chemical Identifiers

10500-11-5
128.171
MRLKTTBPWZXARX-UHFFFAOYSA-N
530323
CCC(C=C)OC(=O)C
C7H12O2
MFCD02093427
Acetic Acid 1-Ethylallyl Ester, Acetic Acid 1-Penten-3-yl Ester, 1-Ethylallyl Acetate
pent-1-en-3-yl acetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0760

TSCA : No

SDS
Documents

Documents

Promotions

Promotions