missing translation for 'onlineSavingsMsg'
Learn More

1-Pentanol 99.0+%, TCI America™

Catalog No. P005525ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P005525ML 25 mL
P0055500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P005525ML Supplier TCI America Supplier No. P005525ML

Chemical Identifiers

CAS 71-41-0
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00002977,MFCD00081734,MFCD01075169
InChI Key AMQJEAYHLZJPGS-UHFFFAOYSA-N
Synonym 1-pentanol, amyl alcohol, n-amyl alcohol, n-pentanol, pentanol, pentyl alcohol, butylcarbinol, 1-pentyl alcohol, amylol, n-butylcarbinol
PubChem CID 6276
ChEBI CHEBI:44884
IUPAC Name pentan-1-ol
SMILES CCCCCO

Specifications

Melting Point -79°C
Color Colorless
Boiling Point 137°C
Quantity 25 mL
UN Number 1105
Formula Weight 88.15
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Pentanol
EINECSNumber (2)-0217
RTECSNumber SB9800000
TSCA Yes