missing translation for 'onlineSavingsMsg'
Learn More

1-Pentanol, 99%, pure

CAS: 71-41-0 | C5H12O | 88.15 g/mol

Supplier:  Thermo Scientific Chemicals 160600025

 View more versions of this product

Catalog No. AC160600025



Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Pentanol
98.5
-78.0°C
137.0°C to 139.0°C
Authentic
Glass Bottle
1.4083 to 1.4103
MFCD00002977,MFCD00081734,MFCD01075169
01, 383
15, 7230
Solubility in water: 22g/L (22°C). Other solubilities: miscible with alcohol, benzene and ether, soluble in most organic solvents
CCCCCO
88.15
4 mPa.s (20°C)
88.15
Pure
71-41-0
100.0
0.8110g/mL
49°C
98.5% min. (GC)
C5H12O
CH3(CH2)4OH
2.5 L
0.811
1-pentanol, amyl alcohol, n-amyl alcohol, n-pentanol, pentanol, pentyl alcohol, butylcarbinol, 1-pentyl alcohol, amylol, n-butylcarbinol
AMQJEAYHLZJPGS-UHFFFAOYSA-N
pentan-1-ol
6276
CHEBI:44884
99%
Liquid

Chemical Identifiers

71-41-0
88.15
AMQJEAYHLZJPGS-UHFFFAOYSA-N
6276
pentan-1-ol
C5H12O
MFCD00002977,MFCD00081734,MFCD01075169
1-pentanol, amyl alcohol, n-amyl alcohol, n-pentanol, pentanol, pentyl alcohol, butylcarbinol, 1-pentyl alcohol, amylol, n-butylcarbinol
CHEBI:44884
CCCCCO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
Flammable liquid and vapor.
Causes serious eye irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
Wear protective glove

GHS Signal Word: Warning

EINECSNumber : 200-752-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only