Learn More
1-Pentanol, 99%, pure
CAS: 71-41-0 | C5H12O | 88.15 g/mol
Supplier: Thermo Scientific Chemicals 160600010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1-Pentanol | |
| 98.5 | |
| -78.0°C | |
| 137.0°C to 139.0°C | |
| Authentic | |
| Glass Bottle | |
| 1.4083 to 1.4103 | |
| MFCD00002977,MFCD00081734,MFCD01075169 | |
| 01, 383 | |
| 15, 7230 | |
| Solubility in water: 22g/L (22°C). Other solubilities: miscible with alcohol, benzene and ether, soluble in most organic solvents | |
| CCCCCO | |
| 88.15 | |
| 4 mPa.s (20°C) | |
| 88.15 | |
| Pure |
| 71-41-0 | |
| 100.0 | |
| 0.8110g/mL | |
| 49°C | |
| 98.5% min. (GC) | |
| C5H12O | |
| CH3(CH2)4OH | |
| 1 L | |
| 0.811 | |
| 1-pentanol, amyl alcohol, n-amyl alcohol, n-pentanol, pentanol, pentyl alcohol, butylcarbinol, 1-pentyl alcohol, amylol, n-butylcarbinol | |
| AMQJEAYHLZJPGS-UHFFFAOYSA-N | |
| pentan-1-ol | |
| 6276 | |
| CHEBI:44884 | |
| 99% | |
| Liquid |
Chemical Identifiers
| 71-41-0 | |
| 88.15 | |
| AMQJEAYHLZJPGS-UHFFFAOYSA-N | |
| 6276 | |
| pentan-1-ol |
| C5H12O | |
| MFCD00002977,MFCD00081734,MFCD01075169 | |
| 1-pentanol, amyl alcohol, n-amyl alcohol, n-pentanol, pentanol, pentyl alcohol, butylcarbinol, 1-pentyl alcohol, amylol, n-butylcarbinol | |
| CHEBI:44884 | |
| CCCCCO |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
Flammable liquid and vapor.
Causes serious eye irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
Wear protective glove
GHS Signal Word: Warning
EINECSNumber : 200-752-1
RUO – Research Use Only