missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Pentanethiol 98.0+%, TCI America™
$109.98 - $922.71
Chemical Identifiers
| CAS | 110-66-7 |
|---|---|
| Molecular Formula | C5H12S |
| Molecular Weight (g/mol) | 104.21 |
| MDL Number | MFCD00004907 |
| InChI Key | ZRKMQKLGEQPLNS-UHFFFAOYSA-N |
| Synonym | 1-pentanethiol, pentyl mercaptan, amyl mercaptan, n-amyl mercaptan, pentanethiol, amyl thioalcohol, pentalarm, amyl sulfhydrate, amyl hydrosulfide, pentylmercaptan |
| PubChem CID | 8067 |
| IUPAC Name | pentane-1-thiol |
| SMILES | CCCCCS |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P005325ML
|
TCI America
P005325ML |
25 mL |
Each for $109.98
|
|
|||||
|
P0053500ML
|
TCI America
P0053500ML |
500 mL |
Each for $922.71
|
|
|||||
Chemical Identifiers
| 110-66-7 | |
| 104.21 | |
| ZRKMQKLGEQPLNS-UHFFFAOYSA-N | |
| 8067 | |
| CCCCCS |
| C5H12S | |
| MFCD00004907 | |
| 1-pentanethiol, pentyl mercaptan, amyl mercaptan, n-amyl mercaptan, pentanethiol, amyl thioalcohol, pentalarm, amyl sulfhydrate, amyl hydrosulfide, pentylmercaptan | |
| pentane-1-thiol |
Specifications
| 110-66-7 | |
| 126°C | |
| MFCD00004907 | |
| 1111 | |
| ZRKMQKLGEQPLNS-UHFFFAOYSA-N | |
| pentane-1-thiol | |
| 8067 | |
| ≥98.0% (GC,T) | |
| 1-Pentanethiol |
| Colorless | |
| C5H12S | |
| 25 mL | |
| 1-pentanethiol, pentyl mercaptan, amyl mercaptan, n-amyl mercaptan, pentanethiol, amyl thioalcohol, pentalarm, amyl sulfhydrate, amyl hydrosulfide, pentylmercaptan | |
| CCCCCS | |
| 104.21 | |
| 104.21 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0464
RTECSNumber : SA3150000
TSCA : Yes