Learn More
1-Pentanethiol, 97%
CAS: 110-66-7 | C5H12S | 104.21 g/mol
$124.29 - $124.29
Chemical Identifiers
| CAS | 110-66-7 |
|---|---|
| Molecular Formula | C5H12S |
| Molecular Weight (g/mol) | 104.21 |
| MDL Number | MFCD00004907 |
| InChI Key | ZRKMQKLGEQPLNS-UHFFFAOYSA-N |
| Synonym | 1-pentanethiol, pentyl mercaptan, amyl mercaptan, n-amyl mercaptan, pentanethiol, amyl thioalcohol, pentalarm, amyl sulfhydrate, amyl hydrosulfide, pentylmercaptan |
| PubChem CID | 8067 |
| IUPAC Name | pentane-1-thiol |
| SMILES | CCCCCS |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0433014
|
Thermo Scientific Chemicals
L0433014 |
25 g |
Each for $124.29
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 110-66-7 | |
| 104.21 | |
| ZRKMQKLGEQPLNS-UHFFFAOYSA-N | |
| 8067 | |
| CCCCCS |
| C5H12S | |
| MFCD00004907 | |
| 1-pentanethiol, pentyl mercaptan, amyl mercaptan, n-amyl mercaptan, pentanethiol, amyl thioalcohol, pentalarm, amyl sulfhydrate, amyl hydrosulfide, pentylmercaptan | |
| pentane-1-thiol |
Specifications
| 110-66-7 | |
| 0.841 | |
| 18°C (64°F) | |
| C5H12S | |
| MFCD00004907 | |
| UN1111 | |
| Air Sensitive | |
| ZRKMQKLGEQPLNS-UHFFFAOYSA-N | |
| pentane-1-thiol | |
| 8067 | |
| 97% |
| -76°C | |
| 125°C to 126°C | |
| Stench | |
| 1.446 | |
| 25 g | |
| 1730979 | |
| 1-pentanethiol, pentyl mercaptan, amyl mercaptan, n-amyl mercaptan, pentanethiol, amyl thioalcohol, pentalarm, amyl sulfhydrate, amyl hydrosulfide, pentylmercaptan | |
| CCCCCS | |
| 104.21 | |
| 104.22 | |
| 1-Pentanethiol |
Safety and Handling
GHS H Statement
H225-H302-H332-H315-H319-H335
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H225-H302+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMYL MERCAPTAN
EINECSNumber : 203-789-1
RTECSNumber : SA3150000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only