missing translation for 'onlineSavingsMsg'
Learn More

1-Pentanesulfonic acid, sodium salt monohydrate, 98+%, HPLC grade

Catalog No. p-179260
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423200250 25 g Glass Bottle
AC423201000 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC423200250 Supplier Thermo Scientific Chemicals Supplier No. 423200250

CAS: 207605-40-1 | C5H11NaO3S·H2O | 192.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ion pair chromatography

Chemical Identifiers

CAS 207605-40-1
Molecular Formula C5H11NaO3S·H2O
Molecular Weight (g/mol) 192.22
MDL Number MFCD00149548
InChI Key FPQYXAFKHLSWTI-UHFFFAOYSA-M
Synonym sodium pentane-1-sulfonate hydrate, 1-pentanesulfonic acid sodium salt monohydrate, sodium 1-pentanesulfonate monohydrate, potassium hydrate pentane-1-sulfonate, sodium 1-pentanesulfonate hydrate, sodium hydrate pentane-1-sulfonate, sodium pentane-1-sulfonate-water 1/1/1, 1-pentane sulphonic acid sodium salt monohydrate, 1-pentanesulfonic acid sodium salt monohydydrate, sodium 1-pentanesulfonate monohydrate, hplc grade
PubChem CID 23693099
IUPAC Name sodium;pentane-1-sulfonate;hydrate
SMILES CCCCCS(=O)(=O)[O-].O.[Na+]

Specifications

Absorbance (0.25M aq. soln.) (1 cm cell vs HPLC water), 0.05 max. at 220nm, 0.03 max. at 230nm, 0.02 max. at 240nm, 0.01 max. at 250nm, 0.01 max. at 260nm
Melting Point >300°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass Bottle
Linear Formula CH3(CH2)4SO3Na·H2O
Quantity 25 g
Beilstein 04, III, 23
Solubility Information Solubility in water: soluble
Formula Weight 192.22
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 1-Pentanesulfonic Acid, Sodium Salt Monohydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
EINECSNumber 245-208-4
TSCA TSCA

RUO – Research Use Only