missing translation for 'onlineSavingsMsg'
Learn More

1-Pentanesulfonic acid, sodium salt hydrate, 99%

CAS: 1266615-65-9 | C5H11NaO3S | 174.19 g/mol

$272.91 - $272.91

Chemical Identifiers

CAS 1266615-65-9
Molecular Formula C5H11NaO3S
Molecular Weight (g/mol) 174.19
MDL Number MFCD00149548
InChI Key ROBLTDOHDSGGDT-UHFFFAOYSA-M
Synonym n-Amylsulfonic acid, sodium salt
IUPAC Name sodium pentane-1-sulfonate
SMILES [Na+].CCCCCS([O-])(=O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC270370250
SDS View Documents
Thermo Scientific Chemicals
270370250
25 g Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1266615-65-9
174.19
ROBLTDOHDSGGDT-UHFFFAOYSA-M
sodium pentane-1-sulfonate
C5H11NaO3S
MFCD00149548
n-Amylsulfonic acid, sodium salt
[Na+].CCCCCS([O-])(=O)=O

Specifications

>300.0°C
White
25 g
C5H11NaO3S
MFCD00149548
n-Amylsulfonic acid, sodium salt
ROBLTDOHDSGGDT-UHFFFAOYSA-M
sodium pentane-1-sulfonate
174.12
Authentic
Glass bottle
1-Pentanesulfonic acid, sodium salt hydrate
1266615-65-9
Crystalline Powder
99%
CH3(CH2)4SO3Na·xH2O
04, III, 23
(10% in water) Clear colorless
[Na+].CCCCCS([O-])(=O)=O
174.19
99%
9.6% max. (100°C, 3 hrs) (vacuum dried)
(10% aq. soln.) clear, colorless
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only