missing translation for 'onlineSavingsMsg'
Learn More

1-Pentanesulfonic Acid Sodium Salt, HPLC Grade, J.T. Baker™

Catalog No. p-5383285
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Wide Mouth Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02002049 25 g Wide Mouth Amber Glass Bottle
02002048 100 g Wide Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02002049 Supplier Avantor Supplier No. 284105
Only null left

High purity solvents for use in analytical and preparative separations

    For Ion-Pair Chromatography of Basic Compounds; For Laboratory,Research,or Manufacturing Use

Chemical Identifiers

CAS 22767-49-3
Molecular Formula C5H11NaO3S
Molecular Weight (g/mol) 174.19
MDL Number MFCD00007541,MFCD00149548
InChI Key ROBLTDOHDSGGDT-UHFFFAOYSA-M
Synonym sodium 1-pentanesulfonate, sodium pentane-1-sulfonate, sodium pentanesulfonate, 1-pentanesulfonic acid sodium salt, 1-pentanesulfonic acid, sodium salt, 1-pentylsodiumsulfonate, ipc-alks-5, 1-pentanesulfonate, sodium, pentanesulfonic acid na-salt, pentanesulfonic acidsodiumsalt
PubChem CID 23664617
IUPAC Name sodium pentane-1-sulfonate
SMILES [Na+].CCCCCS([O-])(=O)=O

Specifications

Packaging Wide Mouth Amber Glass Bottle
Quantity 25 g
Specific Gravity 1L = 0.00kg
Formula Weight 174.21
Grade HPLC
Chemical Name or Material 1-Pentanesulfonic Acid Sodium Salt