missing translation for 'onlineSavingsMsg'
Learn More

1-Pentanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous

Catalog No. AC396650250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC396650250 25 g Glass bottle
AC396651000 100 g Glass bottle
AC396655000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC396650250 Supplier Thermo Scientific Chemicals Supplier No. 396650250

CAS: 22767-49-3 | C5H11NaO3S | 174.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ion pair chromatography

Chemical Identifiers

CAS 22767-49-3
Molecular Formula C5H11NaO3S
Molecular Weight (g/mol) 174.19
MDL Number MFCD00007541,MFCD00149548
InChI Key ROBLTDOHDSGGDT-UHFFFAOYSA-M
Synonym sodium 1-pentanesulfonate, sodium pentane-1-sulfonate, sodium pentanesulfonate, 1-pentanesulfonic acid sodium salt, 1-pentanesulfonic acid, sodium salt, 1-pentylsodiumsulfonate, ipc-alks-5, 1-pentanesulfonate, sodium, pentanesulfonic acid na-salt, pentanesulfonic acidsodiumsalt
PubChem CID 23664617
IUPAC Name sodium;pentane-1-sulfonate
SMILES [Na+].CCCCCS([O-])(=O)=O

Specifications

Absorbance 0.02 max. (C=0.5 M in water) at 260nm, 0.04 max. (C=0.5 M in water) at 230nm, 0.06 max. C=0.5 M in water at 220nm, 0.1 max. (C=0.5 M in water) at 210nm
Melting Point >300.0°C
Color White
Loss on Drying 2% (150°C) max.
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula CH3(CH2)4SO3Na
Quantity 25 g
Solubility Information Solubility in water: soluble.
Formula Weight 174.2
Percent Purity 99+%
Grade Ion Pair Chromatography
Physical Form Crystalline Powder
Chemical Name or Material 1-Pentanesulfonic acid, sodium salt
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
EINECSNumber 245-208-4

RUO – Research Use Only