missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Pentadecanethiol 98.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Boiling Point | 325°C |
| Quantity | 10 mL |
| Formula Weight | 244.48 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystals |
| Chemical Name or Material | 1-Pentadecanethiol |
Safety and Handling
| EINECSNumber | (2)-0464 |
| TSCA | No |