missing translation for 'onlineSavingsMsg'
Learn More

1-Octyne 95.0+%, TCI America™

Catalog No. O005025ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O005025ML 25 mL
O0050100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O005025ML Supplier TCI America Supplier No. O005025ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 629-05-0
Molecular Formula C8H14
Molecular Weight (g/mol) 110.20
MDL Number MFCD00009546
InChI Key UMIPWJGWASORKV-UHFFFAOYSA-N
Synonym 1-octyne, hexylacetylene, octyne-1, n-octyne, hex-1-yl acetylene, pubchem13059, acmc-209na4, 4-01-00-01034 beilstein handbook reference, 1-octyne 10g
PubChem CID 12370
IUPAC Name oct-1-yne
SMILES CCCCCCC#C

Specifications

Color Yellow
Boiling Point 126°C
Quantity 25 mL
UN Number 3295
Formula Weight 110.20
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Octyne
RTECSNumber RI2655000
TSCA Yes