missing translation for 'onlineSavingsMsg'
Learn More

1-Octyne, 99%

CAS: 629-05-0 | C8H14 | 110.20 g/mol

$127.80 - $127.80

Chemical Identifiers

CAS 629-05-0
Molecular Formula C8H14
Molecular Weight (g/mol) 110.20
MDL Number MFCD00009546
InChI Key UMIPWJGWASORKV-UHFFFAOYSA-N
Synonym 1-octyne, hexylacetylene, octyne-1, n-octyne, hex-1-yl acetylene, pubchem13059, acmc-209na4, 4-01-00-01034 beilstein handbook reference, 1-octyne 10g
PubChem CID 12370
IUPAC Name oct-1-yne
SMILES CCCCCCC#C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC223510250
SDS View Documents
Thermo Scientific Chemicals
223510250
25 mL Glass bottle
Each for $127.80
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

629-05-0
110.20
UMIPWJGWASORKV-UHFFFAOYSA-N
12370
CCCCCCC#C
C8H14
MFCD00009546
1-octyne, hexylacetylene, octyne-1, n-octyne, hex-1-yl acetylene, pubchem13059, acmc-209na4, 4-01-00-01034 beilstein handbook reference, 1-octyne 10g
oct-1-yne

Specifications

629-05-0
100.0
Colorless to Yellow
127°C to 128°C
Authentic
Glass bottle
1.4150 to 1.417
MFCD00009546
01, 258
1-octyne, hexylacetylene, octyne-1, n-octyne, hex-1-yl acetylene, pubchem13059, acmc-209na4, 4-01-00-01034 beilstein handbook reference, 1-octyne 10g
UMIPWJGWASORKV-UHFFFAOYSA-N
oct-1-yne
12370
99%
1-Octyne
98.5
-80°C
0.7400g/mL
17°C
99%
C8H14
CH3(CH2)5C∽CH
25 mL
0.74
Solubility in water: 0.024g/L (25°C)
CCCCCCC#C
110.20
110.2
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May be fatal if swallowed and enters airways.
Harmful if swallowed.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 211-069-3

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only