Learn More
1-Octyne, 98%
CAS: 629-05-0 | C8H14 | 110.20 g/mol
Supplier: Thermo Scientific Chemicals A1244014
Description
1-Octyne is widely used in organic reactions such as halogenation, hydration, oxidative cleavage, hydrogenation, hydrohalogenation, nitrile formation, polymerization and substitution reactions. It is used a precursor for the preparation of monohalogen substituted akenes or dihalogen substituted alkanes on treated with hydrogen halides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Octyne is widely used in organic reactions such as halogenation, hydration, oxidative cleavage, hydrogenation, hydrohalogenation, nitrile formation, polymerization and substitution reactions. It is used a precursor for the preparation of monohalogen substituted akenes or dihalogen substituted alkanes on treated with hydrogen halides.
Solubility
Miscible with alcohol, ether and other organic solvents. Immiscible with water.
Notes
Incompatible with oxidizing agents. Keep away from heat, sparks, sources of ignition and flame.
Specifications
| 1-Octyne | |
| -60°C | |
| 123°C to 126°C | |
| 98% | |
| 1.416 | |
| MFCD00009546 | |
| UN3295 | |
| 1-octyne, hexylacetylene, octyne-1, n-octyne, hex-1-yl acetylene, pubchem13059, acmc-209na4, 4-01-00-01034 beilstein handbook reference, 1-octyne 10g | |
| UMIPWJGWASORKV-UHFFFAOYSA-N | |
| oct-1-yne | |
| 12370 | |
| 98% |
| 629-05-0 | |
| 0.746 | |
| 17°C (62°F) | |
| C8H14 | |
| CH3(CH2)5C≡CH | |
| 25 g | |
| 1734494 | |
| Miscible with alcohol,ether and other organic solvents. Immiscible with water. | |
| CCCCCCC#C | |
| 110.20 | |
| 110.2 | |
| Liquid |
Chemical Identifiers
| 629-05-0 | |
| 110.20 | |
| UMIPWJGWASORKV-UHFFFAOYSA-N | |
| 12370 | |
| CCCCCCC#C |
| C8H14 | |
| MFCD00009546 | |
| 1-octyne, hexylacetylene, octyne-1, n-octyne, hex-1-yl acetylene, pubchem13059, acmc-209na4, 4-01-00-01034 beilstein handbook reference, 1-octyne 10g | |
| oct-1-yne |
Safety and Handling
GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
H225-H304
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber : 211-069-3
RTECSNumber : RI2655000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only