missing translation for 'onlineSavingsMsg'
Learn More

1-Octyne, 98%

Catalog No. AAA1244009
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1244009 10 g
AAA1244014 25 g
AAA1244022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1244009 Supplier Thermo Scientific Chemicals Supplier No. A1244009
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 629-05-0 | C8H14 | 110.20 g/mol

1-Octyne is widely used in organic reactions such as halogenation, hydration, oxidative cleavage, hydrogenation, hydrohalogenation, nitrile formation, polymerization and substitution reactions. It is used a precursor for the preparation of monohalogen substituted akenes or dihalogen substituted alkanes on treated with hydrogen halides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octyne is widely used in organic reactions such as halogenation, hydration, oxidative cleavage, hydrogenation, hydrohalogenation, nitrile formation, polymerization and substitution reactions. It is used a precursor for the preparation of monohalogen substituted akenes or dihalogen substituted alkanes on treated with hydrogen halides.

Solubility
Miscible with alcohol, ether and other organic solvents. Immiscible with water.

Notes
Incompatible with oxidizing agents. Keep away from heat, sparks, sources of ignition and flame.

Chemical Identifiers

CAS 629-05-0
Molecular Formula C8H14
Molecular Weight (g/mol) 110.20
MDL Number MFCD00009546
InChI Key UMIPWJGWASORKV-UHFFFAOYSA-N
Synonym 1-octyne, hexylacetylene, octyne-1, n-octyne, hex-1-yl acetylene, pubchem13059, acmc-209na4, 4-01-00-01034 beilstein handbook reference, 1-octyne 10g
PubChem CID 12370
IUPAC Name oct-1-yne
SMILES CCCCCCC#C

Specifications

Melting Point -60°C
Density 0.746
Boiling Point 123°C to 126°C
Flash Point 17°C (62°F)
Assay Percent Range 98%
Refractive Index 1.416
Linear Formula CH3(CH2)5C≡CH
Quantity 10 g
UN Number UN3295
Beilstein 1734494
Solubility Information Miscible with alcohol,ether and other organic solvents. Immiscible with water.
Formula Weight 110.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Octyne
Show More Show Less
Hazard Category H225-H304
Hazard Statement GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 211-069-3
RTECSNumber RI2655000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only