missing translation for 'onlineSavingsMsg'
Learn More

1-Octyn-3-ol 98.0+%, TCI America™

Catalog No. p-7134233
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O019625ML 25 mL
O0196250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O019625ML Supplier TCI America Supplier No. O019625ML

Chemical Identifiers

CAS 818-72-4
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.20
MDL Number MFCD00004588
InChI Key VUGRNZHKYVHZSN-UHFFFAOYNA-N
Synonym 1-octyn-3-ol, 1-octyne-3-ol, oct-1-yne-3-ol, 1-oct-1-yn-3-ol, 3-hydroxyoct-1-yne, acmc-20apfv, acmc-20apkf, rac-1-octyn-3-ol, oct-1 yn-3 ol, pubchem11033
PubChem CID 13166
IUPAC Name oct-1-yn-3-ol
SMILES CCCCCC(O)C#C

Specifications

Color Yellow
Boiling Point 83°C
Quantity 25 mL
Formula Weight 126.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Octyn-3-ol