missing translation for 'onlineSavingsMsg'
Learn More

1-Octylphosphonic dichloride, 97%

Catalog No. p-7047303
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1858706 5 g
AAL1858714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1858706 Supplier Thermo Scientific Chemicals Supplier No. L1858706

CAS: 3095-94-1 | C8H17Cl2OP | 231.097 g/mol

1-Octylphosphonic dichloride is employed in the surface modification of calcium fluoro and hydroxyapatite by 1-octylphosphonic dichloride and qualitative and quantitative analysis of phosphate rock from Hazm Al-jalamid area, northern Saudi Arabia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octylphosphonic dichloride is employed in the surface modification of calcium fluoro and hydroxyapatite by 1-octylphosphonic dichloride and qualitative and quantitative analysis of phosphate rock from Hazm Al-jalamid area, northern Saudi Arabia.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from strong bases, water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Chemical Identifiers

CAS 3095-94-1
Molecular Formula C8H17Cl2OP
Molecular Weight (g/mol) 231.097
MDL Number MFCD00015813
InChI Key RTHCRCDXILKTCY-UHFFFAOYSA-N
Synonym octylphosphonic dichloride, 1-octylphosphonic dichloride, octylphosphonoyl dichloride, acmc-1afrz, octyl phosphonic dichloride, dichloro octyl phosphine oxide, phosphonic dichloride,p-octyl
PubChem CID 76541
IUPAC Name 1-dichlorophosphoryloctane
SMILES CCCCCCCCP(=O)(Cl)Cl

Specifications

Density 1.115
Boiling Point 143°C to 145°C (12 mmHg)
Refractive Index 1.4665
Quantity 5 g
UN Number UN3265
Beilstein 1931202
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 231.1
Percent Purity 97%
Chemical Name or Material 1-Octylphosphonic dichloride
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 221-444-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only