missing translation for 'onlineSavingsMsg'
Learn More

1-Octylphosphonic acid, 98%

Catalog No. p-7049739
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA2065103 1 g
AA2065106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA2065103 Supplier Thermo Scientific Chemicals Supplier No. 02065103
Only null left

CAS: 4724-48-5 | C8H19O3P | 194.211 g/mol

1-Octylphosphonic acid is a basic building block and used in the preparation of various pharmaceutical compounds and chemical products. It is supported with calcium hydroxyapatite and used in the dental field as corrosion inhibitor and organic building block. It acts as a corrosion inhibitor in metallic household surfaces such as faucets and drains as well as prevents the rusting of can or dispenser. It is also used in the synthesis of octyl-phosphonic acid dimethyl ester by reacting with iodomethane in presence of CS2 CO3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4724-48-5
Molecular Formula C8H19O3P
Molecular Weight (g/mol) 194.211
MDL Number MFCD00015841
InChI Key NJGCRMAPOWGWMW-UHFFFAOYSA-N
Synonym n-octylphosphonic acid, 1-octylphosphonic acid, phosphonic acid, octyl, phosphonic acid, p-octyl, unii-95c2aw87bo, octylphosphonicacid, acmc-209uni, p-octyl-phosphonic acid, octylphosphonic acid, dsstox_cid_24679
PubChem CID 78452
IUPAC Name octylphosphonic acid
SMILES CCCCCCCCP(=O)(O)O

Specifications

Melting Point 100°C to 102°C
Quantity 1 g
Beilstein 1764242
Solubility Information Slightly soluble in organic solvents and water.
Formula Weight 194.21
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material 1-Octylphosphonic acid
Hazard Category H302-H314-H335-H373
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
EINECSNumber 225-218-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only