missing translation for 'onlineSavingsMsg'
Learn More

1-Octylboronic acid, 97%

Catalog No. p-7047922
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1996403 1 g
AAL1996406 5 g
AAL1996414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1996403 Supplier Thermo Scientific Chemicals Supplier No. L1996403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 28741-08-4 | C8H19BO2 | 158.048 g/mol

1-Octylboronic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octylboronic acid is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 28741-08-4
Molecular Formula C8H19BO2
Molecular Weight (g/mol) 158.048
MDL Number MFCD01074560
InChI Key GKFRVXOKPXCXAK-UHFFFAOYSA-N
Synonym n-octylboronic acid, 1-octylboronic acid, boronic acid, octyl, 1-n-octaneboronic acid, boronic acid, b-octyl, caprylboronic acid, acmc-1cbf1, n-octaneboronic acid, b-octyl-boronic acid
PubChem CID 5195177
IUPAC Name octylboronic acid
SMILES B(CCCCCCCC)(O)O

Specifications

Melting Point 81°C to 85°C
Quantity 1 g
Beilstein 1739759
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 158.05
Percent Purity 97%
Chemical Name or Material 1-Octylboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only